Learn More
Med Vet International Universal "F" Anesthesia Circuit 60" hose 2 liter, J0248FL
Catalog No. 5000582078
Encompass_Preferred
This is the popular uni-limb circuit that is atube within a tube circuit.Features:• One 40" hose for lighter weight and maneuverability around thepatient’s head.• Exhaled gas warms up incoming fresh gas.• Attaches to any standard anesthesia machine.
Catalog No. 50-005-82078 Supplier Med Vet International Supplier No. JOR248FL
This is the popular uni-limb circuit that is atube within a tube circuit.Features:• One 40" hose for lighter weight and maneuverability around thepatient’s head.• Exhaled gas warms up incoming fresh gas.• Attaches to any standard anesthesia machine.