Learn More
Med Vet International Universal "F" Anesthesia circuit 60" hose w/o bag, J0248FLA
Supplier:  Med Vet International JOR248FLA
Encompass_Preferred
This is the popular uni-limb circuit that is a tube within a tube circuit.Features: One 60" hose for lighter weight and maneuverability around the patient’s head. Exhaled gas warms up incoming fresh gas. Attaches to any standard anesthesia machine.Complete Circuit without 2-liter breathing bag.
Catalog No. 50-005-82984