Learn More
Med Vet International Universal "F" Flex Circuit, w/ Bag 108", J0248W, Each
Catalog No. 5000582373
Encompass_Preferred
The corrugated tubing of this unilimb circuit offers expandable and contraction capability. The clinician can determine the length of the hose needed for every surgery. Better positioning - the hose can be set at varying angles versus just standard straight on tubing. Excess tubing can be contracted and out of the way. Includes right angle elbow with gas port. Flex Circuit. 108" expanded length to 38" contracted length. With 2 liter bag.
Catalog No. 50-005-82373 Supplier Med Vet International Supplier No. JOR248W
The corrugated tubing of this unilimb circuit offers expandable and contraction capability. The clinician can determine the length of the hose needed for every surgery. Better positioning - the hose can be set at varying angles versus just standard straight on tubing. Excess tubing can be contracted and out of the way. Includes right angle elbow with gas port. Flex Circuit. 108" expanded length to 38" contracted length. With 2 liter bag.