Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Universal Fit Pipet Tips: Racked, Sterile
GSA_VAGREENER_CHOICE

Catalog No. 07200301 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Color:
Blue
Natural
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
07-200-301 1 to 200 μL Natural
07-200-304 100 to 1000 μL Blue
07-200-583 1 to 200 μL Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-200-301 Supplier Corning™ Supplier No. 4864
Add to Cart
Add to Cart

Designed to assure a reliable fit with major brand pipetters

  • Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
  • Beveled orifice helps ensure accurate fluid delivery.
  • 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
  • Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
  • RNase-/DNase-free.
  • Nonpyrogenic.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Clean Claims RNase/DNase-free and Nonpyrogenic, DNA free and RNase/DNase free, PCR Certified, Manufactured in a clean room environment
Format Rack
Tip Style Round
Packaging Rack
Quantity 96 Tips/Rack, 10 Racks/Cs., 960 Tips/Cs.
Product Type Universal Fit Pipette Tips
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
Graduations 10 μL, 50 μL and 100 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.