Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Great Lakes Diagnostics™ Universal Fit Pipette Tip

Catalog No. 23444006
Change view
Click to view available options
Volume (Metric):
200 μL
1250 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
23-444-006 1250 μL
23-444-004 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-444-006 Supplier Great Lakes Diagnostics™ Supplier No. FT672201

missing translation for 'validMainframeMsg'

Great Lakes Diagnostics™ universal fit pipette tips are available in different volumes and lengths. Compatible with a broad range of pipettes.

  • Sterile
  • Filtered
  • Racked
  • DNA/RNA free
  • Max Recovery
  • Compatible with:
    • Eppendorf Reference™ SC, vol .5-10 μL, 2-20 μL, 10-100 μL, 50-200 μL, 100-1000 μL
    • Thermo Finnpipette™ F1/F2 SC, 1-10 μL, 2-20 μL, 10-100 μL, 20-200 μL, 100-1000 μL
    • Finnpipette F3 SC 1-10 μL, 10-100 μL
    • Biohit Proline™ SC .5-10 μL, 20 μL, 5-50 μL, 20-200 μL, 100-1000 μL
    • Gilson Pipetman™ SC .5-10 μL, 20 μL, 5-50 μL, 20-200 μL, 100-1000 μL
    • Axygen™ SC and MC pipettes.
    • Corning™ pipettes.

Specifications

Volume (Metric) 1250 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Format Rack
Disposable Single-use
For Use With (Equipment) Axygen SC and MC, Biohit Proline SC, Corning, Eppendorf Reference, Gilson Pipetman SC, Axygen SC and MC, Corning
Graduated No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.