Learn More
MSC OccuNomix Flame Resistant Cap, Nomex, Universal, Double Layer, 1 ea
Catalog No. NC2063784
Encompass
This flame resistant cap offers protection and warmth. It features a universal size and double-layer construction, designed for environments requiring enhanced safety.
  • Flame-resistant
  • Double layer construction for added warmth and protection
  • Slip-on closure
  • Meets ASTM F1506
  • Arc rating ATPV = 7.8 cal/cm²
  • NFPA 70E/CAT 1 compliant
  • Constructed from 8.75 oz Nomex® rib knit material
This item is not returnable. View return policy
Catalog No. NC2063784 Supplier MSC Supplier No. 96553292
This flame resistant cap offers protection and warmth. It features a universal size and double-layer construction, designed for environments requiring enhanced safety.
  • Flame-resistant
  • Double layer construction for added warmth and protection
  • Slip-on closure
  • Meets ASTM F1506
  • Arc rating ATPV = 7.8 cal/cm²
  • NFPA 70E/CAT 1 compliant
  • Constructed from 8.75 oz Nomex® rib knit material