Learn More
Research Products International Corp POXYGRID MICRO-PIPETTOR RACK, UNIVERSAL FREE STANDING
SDP
Catalog No. 50212959
Encompass
These racks keep micro-pipettors neatly organized for quick easy access. Racks will accommodate Pipetman, Finnpipette, Socorex, Eppendorf and most other popular micro-pipettors. Racks hold up to 10 pipettors in two stepped rows of 5 each. End holes fit the Drummond Pipette-Aid pipette filler. A lower shelf holds the tapered ends of the pipettes vertical to prevent barrel damage. Racks are constructed of epoxy coated aluminum that is corrosion resistant.
WARNING  This product can expose you to chemicals including Titanium Dioxide, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-212-959 Supplier Research Products International Corp Supplier No. H189620000
These racks keep micro-pipettors neatly organized for quick easy access. Racks will accommodate Pipetman, Finnpipette, Socorex, Eppendorf and most other popular micro-pipettors. Racks hold up to 10 pipettors in two stepped rows of 5 each. End holes fit the Drummond Pipette-Aid pipette filler. A lower shelf holds the tapered ends of the pipettes vertical to prevent barrel damage. Racks are constructed of epoxy coated aluminum that is corrosion resistant.
WARNING  This product can expose you to chemicals including Titanium Dioxide, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.