Learn More
Biocare Medical, LLC 4plus Biotinylated Universal Goat Link - 100 ml
SDP
Catalog No. 5082872
Encompass_Preferred
This biotinylated antibody reacts with heavy and light chains on mouse and rabbit IgG. It reacts with all mouse IgG subclasses and mouse IgM. It has been affinity-purified with minimal cross-reaction to human, bovine and horse serum proteins. A long spacer arm has been positioned between the biotin and the secondary antibody, which provides a significant increase in sensitivity when coupled with streptavidin conjugates. Biocare Universal Link can be used with either streptavidin horseradish peroxidase or streptavidin alkaline phosphatase detection systems.
Catalog No. 50-828-72 Supplier Biocare Medical, LLC Supplier No. GU600L
This biotinylated antibody reacts with heavy and light chains on mouse and rabbit IgG. It reacts with all mouse IgG subclasses and mouse IgM. It has been affinity-purified with minimal cross-reaction to human, bovine and horse serum proteins. A long spacer arm has been positioned between the biotin and the secondary antibody, which provides a significant increase in sensitivity when coupled with streptavidin conjugates. Biocare Universal Link can be used with either streptavidin horseradish peroxidase or streptavidin alkaline phosphatase detection systems.