Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bolle Safety Universal Goggle Safety Pc Clear Platinum

Catalog No. 17020380
Change view
Click to view available options
Lens Coating:
Platinum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating
17-020-380 Platinum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-020-380 Supplier Bolle Safety Supplier No. UNIVGN12W
Add to Cart
Add to Cart

Universal Goggle Safety Pc Clear Platinum - Pc/Tpr Top Bottom Indirect Vented With Foam and Faceguard

  • The Universal Goggle provides optimum protection and comfort in all circumstances thanks to its ergonomic design.
  • This innovative goggle with its exclusive Platinum coating is available in several versions, each adapted to a specific work environment to offer more peace of mind and efficiency for the worker.
  • Equipped with 'Wave' technology, the Universal Goggle adapts perfectly to each user's morphology, eliminating gaps without increasing pressure points, thus guaranteeing superior protection without compromising the user's comfort.
  • To improve the environmental impact of the product, it has been specifically designed to be easier to recycle: it can be quickly disassembled to separate the materials from each other.

Specifications

Antigen CD20
Width (English) 12.2 in.
Width (Metric) 31 cm
Height (Metric) 3.5 cm
Shape Slightly curved
Lens Coating Platinum
Height (English) 1.38 in.
Certifications/Compliance Crown Z87+ D3 D4 / Visor : Crown Z87+
Applications Flow Cytometry
Classification Monoclonal
Color Yellow
Clone QCH6A7
Concentration 0.2 mg/mL
Conjugate APC
Lens Tint Clear
Formulation PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Product Type Safety Goggles, Antibody
Gene Ms4a1
For Use With (Equipment) Safety and ventilation equipment
Gene Accession No. P19437
Dimensions (L x W x H) 11.8 x 12.2 x 1.38 in.
For Use With (Application) Safety goggles for enviroments needing ventilation
Gene Symbols Ms4a1
Frame Color Yellow
Host Species Rat
Headband Material Polyester, Spandex
Includes Goggle, strap
Length (English) 11.8 in.
Purification Method Affinity chromatography
Length (Metric) 30 cm
Quantity 25 μg
Regulatory Status RUO
Model Universal
Primary or Secondary Primary
Gene ID (Entrez) 12482
Temple Style Polyester strap
Style Adaptability and Pioneer
Weight (English) 0.365967 lb.
Target Species Mouse
Ventilation Details Vented goggles
Content And Storage 4° C, store in dark, DO NOT FREEZE!
Weight (Metric) 166 g
Material Polycarbonate, TPR
Form Liquid
Disposable Reusable
Isotype IgG2a κ
Lens Material Polycarbonate
Frame Material Polycarbonate, TPR
UV Protection None
Frame Type Classic
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.