Learn More
Velp™ Universal H-Stand
SDP
Catalog No. 11392127
Change view
Click to view available options
Dimensions (L x W x H):
462 x 430 mm
Catalog No. Dimensions (L x W x H)
11-392-127 462 x 430 mm
1 options
This item is not returnable. View return policy
Catalog No. 11-392-127 Supplier Velp™ Supplier No. A00000366
Add to Cart
Add to Cart
This item is not returnable. View return policy
Universal H-Stand ensures stability and compatibility with all VELP™ Overhead Stirrers.
  • Great solution for using overhead stirrers on laboratory benches
  • Maximum stability thanks to H-shape of the base
  • Maximum resistance with premium materials such as aluminum and stainless steel, properly treated to resist chemicals and corrosion
  • Possible to adjust stand in order to adapt it to uneven surface
Specifications
Type Stirrer Stand
Dimensions (L x W x H) 462 x 430 mm
Coating Uncoated
Material Stainless Steel