Learn More
Abcam Universal HIER antigen retrieval reagent (10 x ), 100ML
Catalog No. NC1774703
Encompass_Preferred
Universal HIER antigen retrieval reagent (10X) is a heat-induced epitope retrieval (HIER) solution that is compatible with virtually all antibodies and eliminates the need for multiple buffers, including citrate buffer, EDTA or high pH Tris buffers. - Cited in over 20 publications - Colour-coded pH indicator for accuracy - Superior pH stability at high temperatures - Non-toxic, odorless, and azide-free - Optimized for FFPE tissue antigen retrieval
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1774703 Supplier Abcam Supplier No. AB208572100ML
Universal HIER antigen retrieval reagent (10X) is a heat-induced epitope retrieval (HIER) solution that is compatible with virtually all antibodies and eliminates the need for multiple buffers, including citrate buffer, EDTA or high pH Tris buffers. - Cited in over 20 publications - Colour-coded pH indicator for accuracy - Superior pH stability at high temperatures - Non-toxic, odorless, and azide-free - Optimized for FFPE tissue antigen retrieval
The product is subject to the following: Abcam Restricted Use Statement