Learn More
Agilent Technologies UNIVERSAL HUMAN REFERENCE RNA
Catalog No. 50126134
Encompass
Agilent's Universal Human Reference RNA is composed of total RNA from 10 human cell lines. The reference RNA is designed to be used as a reference for uarray gene-profiling experiments.
Catalog No. 50-126-134 Supplier Agilent Technologies Supplier No. 740000
Agilent's Universal Human Reference RNA is composed of total RNA from 10 human cell lines. The reference RNA is designed to be used as a reference for uarray gene-profiling experiments.