Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Universal Hydrometer
SCI_ED

Catalog No. S30449
Change view
Click to view available options
For Use With (Application):
Measure the specific gravity
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S30449 Measure the specific gravity
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30449 Supplier G&W Instruments Inc Supplier No. A1532

Ideal for specific gravity experiments.

  • Specific gravity range: 0.700-2.000, measured in 0.010 intervals
  • 14.9 in. (38cm) long with a metal ballast

Specifications

Increments 0.01
Length (Metric) 380 mm
Includes Metal ballast
For Use With (Application) Measure the specific gravity
Specific Gravity Range 0.7 to 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.