Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Universal Indicator Solution, Lab Grade, Innovating Science™
SCI_ED

Catalog No. S02412 Shop All Aldon Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02412 1 L
S02409 500 mL
S02410 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
WARNING! :This product can expose you to Phenolphthalein, Methanol and Methyl isobutyl ketone, which are known to the State of California to cause cancer and birth defects or other reproductive harm.
Catalog No. S02412 Supplier Aldon Corporation Supplier No. IS30001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING! :This product can expose you to Phenolphthalein, Methanol and Methyl isobutyl ketone, which are known to the State of California to cause cancer and birth defects or other reproductive harm.

Specifications

Chemical Name or Material Universal pH Indicator
Boiling Point 75 to 80°C
CAS 64-17-5
Grade Laboratory
Packaging HDPE
Color Dark Green
pH Approx. 7
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.