Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Universal Linear Pipette Stand
GREENER_CHOICE

Catalog No. 14223583 Shop All Axygen Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-223-583 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-223-583 Supplier Axygen™ Supplier No. APUSD4
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Convenient pipette storage

Axygen™ universal linear stand for four pipettors, (four single-channel, maximum of two 8-channel or 12-channel pipettors)

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Holds Single-channel Pipettes (4) or Multi-channel Pipettes (2)
Positions 4
For Use With (Application) Liquid Handling
For Use With (Equipment) Pipettes
Product Type Pipette Stand
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.