Learn More
Poltex Universal Locking Wipe Cannister Holder
SDP
Catalog No. 22000515
Change view
Click to view available options
Mounting:
Magnetic Mount
Wall Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-000-515 Wall Mount
22-000-514 Magnetic Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-000-515 Supplier Poltex Supplier No. MINMAX59W
Add to Cart
Add to Cart
Holds virtually any wipe cannister, mask box, glove box, tissue box, etc.
  • Locks securely with padlock (included)
  • Comes with powerful magnets for mounting
  • Inside height adjusts in 0.5 in. increments from 5 to 9 in.
  • Choose from keyhole slots for wall (or countertop) mount (included)
  • Constructed from durable and cleanable 0.25 in. thick PETG
  • Holds virtually and wipe cannister, mask box, glove box, tissue box, wipe packet, etc.
  • Note:
    • Consumables not included
  • Specifications
    Product Type Adjustable, Lockable Dispenser
    Mounting Wall Mount