Learn More
Zymo Research Corporation Universal Methylated Mouse DNA Standard
SDP
Catalog No. 50444308
Encompass_Preferred
The primer set is designed to amplify a fragment of the mouse MLH1 mismatch repair gene following bisulfite treatment. The methylated cytosines comprising CG dinucleotides remain unconverted following bisulfite treatment, whereas non-methylated cytosines are converted into uracil and detected as thymine after PCR.
Catalog No. 50-444-308 Supplier Zymo Research Corporation Supplier No. D5012
The primer set is designed to amplify a fragment of the mouse MLH1 mismatch repair gene following bisulfite treatment. The methylated cytosines comprising CG dinucleotides remain unconverted following bisulfite treatment, whereas non-methylated cytosines are converted into uracil and detected as thymine after PCR.