Learn More
Glas Col LLC Shaker Accessory
SDP
Supplier:  Glas Col LLC 099A BT6000
Encompass_Preferred
Universal mounting platform for 099A S60012 shaker. Required for using 099A BT1000S and 099A BT2000S seperatory funnel holders.
Catalog No. NC1649080