Learn More
Agilent Technologies UNIVERSAL MOUSE REFERENCE RNA
Catalog No. 50126135
Encompass
Universal Mouse Reference RNA 400 ugs 2 tubes x 200microgram each RNase-free water 1.5mL High-quality total RNA for mouse microarray gene-expression profiling Acts as a consistent control for standard data set comparisons
Catalog No. 50-126-135 Supplier Agilent Technologies Supplier No. 740100
Universal Mouse Reference RNA 400 ugs 2 tubes x 200microgram each RNase-free water 1.5mL High-quality total RNA for mouse microarray gene-expression profiling Acts as a consistent control for standard data set comparisons