Learn More
ATCC Universal Mycoplasma Detection Kit (ATCC 30-1012K™)
Catalog No. 502383275
Encompass
The Universal Mycoplasma Detection Kit (ATCC 30-1012K™) is a PCR-based assay designed to rapidly and reliably detect mycoplasma contamination in cell cultures. It includes a lysis buffer and all necessary reagents and primers targeting the 16S rRNA region of over 120 species of Mycoplasma, Acholeplasma, Spiroplasma, and Ureaplasma, including the species most likely to afflict cell cultures. This product is recommended for routine QC in cell culture labs.
Catalog No. 50-238-3275 Supplier ATCC Supplier No. 301012K
The Universal Mycoplasma Detection Kit (ATCC 30-1012K™) is a PCR-based assay designed to rapidly and reliably detect mycoplasma contamination in cell cultures. It includes a lysis buffer and all necessary reagents and primers targeting the 16S rRNA region of over 120 species of Mycoplasma, Acholeplasma, Spiroplasma, and Ureaplasma, including the species most likely to afflict cell cultures. This product is recommended for routine QC in cell culture labs.