Learn More
Alkali Scientific Universal Nasopharyngeal Swab kit for COVID 19 and other virus, room temp storage 20 days, sterile, includes swabs, 2.0mL ITM transport media & 5.0mL vials, kit of 50, 50/Pk
SDP
Catalog No. 5000662458
Encompass_Preferred
Universal Nasopharyngeal Swab Kit designed for reliable specimen collection and transport during COVID 19 and other viral testing procedures. Each sterile kit includes high quality nasopharyngeal swabs, 2.0 mL ITM viral transport media, and durable 5.0 mL transport vials to help preserve sample integrity during handling and shipping. The transport media supports stabilization of viral samples for consistent downstream diagnostic analysis. Components are individually prepared to maintain sterility and minimize contamination risk in clinical and laboratory environments. The kit allows convenient room temperature storage for up to 20 days, helping simplify logistics for sample collection and transport. Ideal for hospitals, diagnostic labs, and testing centers performing viral detection protocols. Supplied as a complete kit of 50 units per pack for efficient laboratory workflow and testing readiness.
Catalog No. 50-006-62458 Supplier Alkali Scientific Supplier No. A2282
Universal Nasopharyngeal Swab Kit designed for reliable specimen collection and transport during COVID 19 and other viral testing procedures. Each sterile kit includes high quality nasopharyngeal swabs, 2.0 mL ITM viral transport media, and durable 5.0 mL transport vials to help preserve sample integrity during handling and shipping. The transport media supports stabilization of viral samples for consistent downstream diagnostic analysis. Components are individually prepared to maintain sterility and minimize contamination risk in clinical and laboratory environments. The kit allows convenient room temperature storage for up to 20 days, helping simplify logistics for sample collection and transport. Ideal for hospitals, diagnostic labs, and testing centers performing viral detection protocols. Supplied as a complete kit of 50 units per pack for efficient laboratory workflow and testing readiness.