Learn More
Alkali Scientific Universal Nasopharyngeal Swab kit for COVID 19 and other virus
SDP
Catalog No. 5000662668
Encompass_Preferred
Universal Nasopharyngeal Swab Kit designed for the safe collection, transport, and short term storage of clinical specimens for COVID 19 and other respiratory virus testing. Each sterile kit includes high quality nasopharyngeal swabs and 3 mL of ITM transport media contained within durable 10 mL screw cap vials to help maintain specimen stability during handling and transport. The transport medium supports viral preservation and is suitable for laboratory diagnostic workflows and molecular testing applications. Components are manufactured under strict quality standards and packaged sterile to minimize contamination risk. The kit allows convenient room temperature storage for up to 20 days, supporting flexible logistics for sample collection and shipping. Supplied as a complete ready to use kit of 100 units per pack, ideal for hospitals, clinics, laboratories, and testing facilities requiring reliable specimen collection materials.
Catalog No. 50-006-62668 Supplier Alkali Scientific Supplier No. A2283
Universal Nasopharyngeal Swab Kit designed for the safe collection, transport, and short term storage of clinical specimens for COVID 19 and other respiratory virus testing. Each sterile kit includes high quality nasopharyngeal swabs and 3 mL of ITM transport media contained within durable 10 mL screw cap vials to help maintain specimen stability during handling and transport. The transport medium supports viral preservation and is suitable for laboratory diagnostic workflows and molecular testing applications. Components are manufactured under strict quality standards and packaged sterile to minimize contamination risk. The kit allows convenient room temperature storage for up to 20 days, supporting flexible logistics for sample collection and shipping. Supplied as a complete ready to use kit of 100 units per pack, ideal for hospitals, clinics, laboratories, and testing facilities requiring reliable specimen collection materials.