Learn More
Alkali Scientific Universal Nasopharyngeal Swab Kit for COVID 19 & Viruses
SDP
Catalog No. 5000662589
Encompass_Preferred
Universal Nasopharyngeal Swab Kit designed for the collection, preservation, and transport of respiratory virus specimens including COVID 19 and other viral pathogens. The kit supports reliable sampling for diagnostic and research workflows. Samples are stabilized in viral transport media (VTM) and recommended to be stored or transported at approximately 4 °C to maintain specimen integrity during handling and shipment. Each sterile kit includes high quality nasopharyngeal swabs, 2.5 mL viral transport media, and durable 5.0 mL transport vials for secure containment. Components are individually packaged to maintain sterility and reduce contamination risk. Suitable for clinical laboratories, testing facilities, and research environments requiring consistent viral specimen collection and transport. Packaged as a kit of 100, single packed units, 100 per pack.
Catalog No. 50-006-62589 Supplier Alkali Scientific Supplier No. A2246
Universal Nasopharyngeal Swab Kit designed for the collection, preservation, and transport of respiratory virus specimens including COVID 19 and other viral pathogens. The kit supports reliable sampling for diagnostic and research workflows. Samples are stabilized in viral transport media (VTM) and recommended to be stored or transported at approximately 4 °C to maintain specimen integrity during handling and shipment. Each sterile kit includes high quality nasopharyngeal swabs, 2.5 mL viral transport media, and durable 5.0 mL transport vials for secure containment. Components are individually packaged to maintain sterility and reduce contamination risk. Suitable for clinical laboratories, testing facilities, and research environments requiring consistent viral specimen collection and transport. Packaged as a kit of 100, single packed units, 100 per pack.