Learn More
Biocare Medical, LLC Universal Negative Control Serum - 6.0 ml
SDP
Catalog No. 5082965
Encompass_Preferred
The Universal Negative Control Serum can be used with any of Biocare mouse and/or rabbit streptavidin kits. It is intended for use as a negative control for either mouse or rabbit antibodies. The Universal Negative Control Serum has been titered for Biocare's monoclonal and polyclonal antibodies, and antibody cocktails. It can also be used on an automated staining system such as the intelliPATH. For all of Biocare's polymer detection systems please use NC499, Polymer Negative Control Serum.
Catalog No. 50-829-65 Supplier Biocare Medical, LLC Supplier No. NC498AA
The Universal Negative Control Serum can be used with any of Biocare mouse and/or rabbit streptavidin kits. It is intended for use as a negative control for either mouse or rabbit antibodies. The Universal Negative Control Serum has been titered for Biocare's monoclonal and polyclonal antibodies, and antibody cocktails. It can also be used on an automated staining system such as the intelliPATH. For all of Biocare's polymer detection systems please use NC499, Polymer Negative Control Serum.