Learn More
TA Instruments Universal Optical Accessory Kit, Provides Interface for Near IR or Raman Spectrometer Probe, Compatible with Discovery DSC Instruments, Includes Silver Lids Quartz Window Adapter Base and Covers
Catalog No. 5000580684 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Universal Optical Accessory Kit provides an interface for installation of a near IR or Raman spectrometer probe. It is compatible with Discovery DSC instruments except Discovery X3. The kit includes inner and outer DSC silver lids, UV quartz window, adapter base, manual lids, and covers. An adaptor ring for Discovery Series DSC is also included. Probe adapter must be supplied by the spectrometer vendor.
Catalog No. 50-005-80684 Supplier TA Instruments Supplier No. 935051.901
Waters TA Instruments Universal Optical Accessory Kit provides an interface for installation of a near IR or Raman spectrometer probe. It is compatible with Discovery DSC instruments except Discovery X3. The kit includes inner and outer DSC silver lids, UV quartz window, adapter base, manual lids, and covers. An adaptor ring for Discovery Series DSC is also included. Probe adapter must be supplied by the spectrometer vendor.