Learn More
Alkali Scientific Universal Oropharyngeal Swab kit for COVID 19 and other virus, room temp storage 20 days, sterile includes swabs, 2.0mL ITM transport media & 5.0mL vials, kit of 50, 50/Pk
SDP
Catalog No. 5000662662
Encompass_Preferred
Universal Oropharyngeal Swab Kit designed for the collection and transport of samples for COVID 19 and other viral testing applications. The kit supports reliable specimen handling with room temperature stability for up to 20 days, making it suitable for laboratories, clinics, and diagnostic testing environments. Each sterile kit includes individually wrapped oropharyngeal swabs and 5.0 mL externally threaded vials pre filled with 2.0 mL ITM transport medium to preserve sample integrity during transport and storage. The secure threaded design helps prevent leakage while maintaining sterility. Components are packaged for convenience and efficient sample processing. Each pack contains 50 sterile vials and 50 swabs, organized as 10 vials per pack and supplied as a complete kit of 50, ideal for routine viral sampling workflows and clinical diagnostic testing procedures.
Catalog No. 50-006-62662 Supplier Alkali Scientific Supplier No. A2280
Universal Oropharyngeal Swab Kit designed for the collection and transport of samples for COVID 19 and other viral testing applications. The kit supports reliable specimen handling with room temperature stability for up to 20 days, making it suitable for laboratories, clinics, and diagnostic testing environments. Each sterile kit includes individually wrapped oropharyngeal swabs and 5.0 mL externally threaded vials pre filled with 2.0 mL ITM transport medium to preserve sample integrity during transport and storage. The secure threaded design helps prevent leakage while maintaining sterility. Components are packaged for convenience and efficient sample processing. Each pack contains 50 sterile vials and 50 swabs, organized as 10 vials per pack and supplied as a complete kit of 50, ideal for routine viral sampling workflows and clinical diagnostic testing procedures.