Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Universal Pipet Tips

Catalog No. 22170173
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Color:
Blue
Natural
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
22-170-173 1 to 200 μL Natural
22-170-403 100 to 1000 μL Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-170-173 Supplier Globe Scientific Supplier No. 151150
Add to Cart
Add to Cart

Available as non-sterile

  • Made from prime virgin polypropylene
  • Universal fit, used with many pipets
  • Reference tips feature graduation marks for precise measurements
  • Autoclavable (121°C at 15 minutes)

Specifications

Volume (Metric) 1 to 200 μL
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Length (Metric) 54mm
Certifications/Compliance Certified free from Human DNA, RNase, DNase, Pyrogens, PCR Inhibitors
Clean Claims DNase, RNase, and Pyrogen free
For Use With (Application) Pipetting
Format Bulk
Tip Style Universal
Packaging Bulk Bag of 1000
Quantity 1000/Bag
Product Type Certified Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
Graduations 10, 50, 100μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.