Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Universal Pipet Tips

Catalog No. 22170403 Shop All Globe Scientific Products
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Color:
Blue
Natural
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
22-170-403 100 to 1000 μL Blue
22-170-173 1 to 200 μL Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-170-403 Supplier Globe Scientific Supplier No. 151146
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Available as non-sterile

  • Made from prime virgin polypropylene
  • Universal fit, used with many pipets
  • Reference tips feature graduation marks for precise measurements
  • Autoclavable (121°C at 15 minutes)

Specifications

Volume (Metric) 100 to 1000 μL
Sterility Non-sterile
Color Blue
Autoclavable Autoclavable
Format Bulk
Tip Style Universal
Product Type Pipette Tip
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.