Learn More
Medicus Health Universal Pipette Stand
SDP
Supplier:  Medicus Health 7615M1
Encompass
• Universal Pipette Stand positions pipettes so that they are easy to identify and reach
• Stands are affordable yet sleek stands and better organization to your liquid handling devices
• Universal Pipette Stand is easy to assemble and are very sturdy unlike may flimsy plastic pipette racks
• Keep your valuable liquid handling tools readily accessible with this adaptable and user-friendly pipette rack
• Offers a secure and convenient storage solution for pipettes and accommodates single-channel, multi-channel and electronic pipettes
• Designed to accommodate pipettes from various manufacturers and sizes with ease
• Crafted from durable, chemical-resistant materials
• Compact design occupies less bench space
• Made from durable ABS
• Dimensions: 14”W x 6”D x 12”H
Catalog No. NC3886391