Learn More
Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
GSA_VA
Catalog No. 11676017 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Flask Clamp
Flask Clamp Kit
Microplate Clamp
Holds:
1 L Erlenmeyer Flask
125 mL Erlenmeyer Flask
2 L Erlenmeyer Flask
25 mL Erlenmeyer Flask
250/300 mL Erlenmeyer Flask
2800 mL Fernbach Flask
4 L Erlenmeyer Flask
5 L Erlenmeyer Flask
50 mL Erlenmeyer Flask
500 mL Erlenmeyer Flask
6 L Erlenmeyer Flask
Flasks
Microplates
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Holds
11-676-017 Flask Clamp 2 L Erlenmeyer Flask
11-676-011 Flask Clamp 25 mL Erlenmeyer Flask
11-676-012 Flask Clamp 50 mL Erlenmeyer Flask
11-676-013 Flask Clamp 125 mL Erlenmeyer Flask
11-676-014 Flask Clamp 250/300 mL Erlenmeyer Flask
11-676-015 Flask Clamp 500 mL Erlenmeyer Flask
11-676-016 Flask Clamp 1 L Erlenmeyer Flask
11-676-018 Flask Clamp 2800 mL Fernbach Flask
11-676-019 Microplate Clamp Microplates
236017 Flask Clamp 4 L Erlenmeyer Flask
236018 Flask Clamp 6 L Erlenmeyer Flask
11-676-020 Flask Clamp Kit Flasks
236028 Flask Clamp 5 L Erlenmeyer Flask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 11-676-017 Supplier Thermo Scientific™ Supplier No. 236016
  • $228.50
    $152.65 / Each of 1
    Save $75.85
    33% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
Secure vessels using these universal platform clamps for orbital shakers.
Secure vessels using Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers. With exceptional innovation, reliability and accuracy, these orbital shakers, platforms and accessories are the optimal choice for a busy lab.
Flask Clamp Starter Kit includes:
  • (1) microplate clamp
  • (2) 25mL flask clamps
  • (2) 50mL flask clamps
  • (2) 125mL flask clamps
  • (4) 250/300mL flask clamps
  • (4) 500mL flask clamps
  • (2) 1L flask clamps
  • (2) 2L flask clamps
  • Replacement screws
Note:
Replacement screws also available separately.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Holds 2 L Erlenmeyer Flask
Product Type Flask Clamp
For Use With (Equipment) MaxQ™ 8000 Shaker