Learn More
Agilent Technologies UNIVERSAL RAT REFERENCE RNA
Catalog No. 50126136
Encompass
Universal Rat Reference RNA, High-quality, pooled total RNA for rat microarray experiments, Acts as a consistent control for Standard data set comparisons, Comprised of 14 different cell lines for broad gene coverage
Catalog No. 50-126-136 Supplier Agilent Technologies Supplier No. 740200
Universal Rat Reference RNA, High-quality, pooled total RNA for rat microarray experiments, Acts as a consistent control for Standard data set comparisons, Comprised of 14 different cell lines for broad gene coverage