Learn More
Biochain Institute Inc Universal RNA - Chicken Normal Tissues, 2x100 ug/PK
SDP
Catalog No. 50184234
Encompass_Preferred
BioChain's Universal RNA is the total RNA prepared with diversified tissues to ensure the broadest coverage of genes. It can be used as controls for microarray study. The Universal RNA is made on an industrial scale to minimize the lot-to-lot variation.
Catalog No. 50-184-234 Supplier Biochain Institute Inc Supplier No. R4C345651
BioChain's Universal RNA is the total RNA prepared with diversified tissues to ensure the broadest coverage of genes. It can be used as controls for microarray study. The Universal RNA is made on an industrial scale to minimize the lot-to-lot variation.