Learn More
Bioworld Universal RNA Dye, 1.5 mL
Catalog No. 50196797
Encompass_Preferred
Premixed and ready-to-load gel dyes.Glow Loading Dyes generate intense DNA/RNA bands with little background or band distortion and can be used with any type of agarose or acrylamide gels. RNA Loading Dyes use formamide and are ideal for quick screenings of RNA integrity. Universal RNA dye contains xylene cyanol in a ficoll-based solution with formamide and SDS (6X concentration).
Catalog No. 50-196-797 Supplier Bioworld Supplier No. 105700291
Premixed and ready-to-load gel dyes.Glow Loading Dyes generate intense DNA/RNA bands with little background or band distortion and can be used with any type of agarose or acrylamide gels. RNA Loading Dyes use formamide and are ideal for quick screenings of RNA integrity. Universal RNA dye contains xylene cyanol in a ficoll-based solution with formamide and SDS (6X concentration).