Learn More
Biochain Institute Inc Universal RNA - Human Normal Tissues, 4x100 ug/PK
SDP
Catalog No. 50184237
Encompass_Preferred
BioChain's Universal RNA is the total RNA prepared with diversified tissues to ensure the broadest coverage of genes. It can be used as controls for microarray study. The Universal RNA is made on an industrial scale to minimize the lot-to-lot variation.
Catalog No. 50-184-237 Supplier Biochain Institute Inc Supplier No. R4234565
BioChain's Universal RNA is the total RNA prepared with diversified tissues to ensure the broadest coverage of genes. It can be used as controls for microarray study. The Universal RNA is made on an industrial scale to minimize the lot-to-lot variation.