Learn More
Biochain Institute Inc Universal RNA - Human Normal Tissues, 2x100 ug/PK
SDP
Catalog No. 50184238
Encompass_Preferred
BioChain's Universal RNA is the total RNA prepared with diversified tissues to ensure the broadest coverage of genes. It can be used as controls for microarray study. The Universal RNA is made on an industrial scale to minimize the lot-to-lot variation.
Catalog No. 50-184-238 Supplier Biochain Institute Inc Supplier No. R42345651
BioChain's Universal RNA is the total RNA prepared with diversified tissues to ensure the broadest coverage of genes. It can be used as controls for microarray study. The Universal RNA is made on an industrial scale to minimize the lot-to-lot variation.