Learn More
Biochain Institute Inc Universal RNA - Mouse Normal Tissues, 4x100 ug/PK
SDP
Catalog No. 50184241
Encompass_Preferred
BioChain's Universal RNA is the total RNA prepared with diversified tissues to ensure the broadest coverage of genes. It can be used as controls for microarray study. The Universal RNA is made on an industrial scale to minimize the lot-to-lot variation.
Catalog No. 50-184-241 Supplier Biochain Institute Inc Supplier No. R4334566
BioChain's Universal RNA is the total RNA prepared with diversified tissues to ensure the broadest coverage of genes. It can be used as controls for microarray study. The Universal RNA is made on an industrial scale to minimize the lot-to-lot variation.