Learn More
Biochain Institute Inc Universal RNA - Rat Normal Tissues, 2x100 ug/PK
SDP
Catalog No. 50184244
Encompass_Preferred
BioChain's Universal RNA is the total RNA prepared with diversified tissues to ensure the broadest coverage of genes. It can be used as controls for microarray study. The Universal RNA is made on an industrial scale to minimize the lot-to-lot variation.
Catalog No. 50-184-244 Supplier Biochain Institute Inc Supplier No. R44345671
BioChain's Universal RNA is the total RNA prepared with diversified tissues to ensure the broadest coverage of genes. It can be used as controls for microarray study. The Universal RNA is made on an industrial scale to minimize the lot-to-lot variation.