Learn More
Brady Universal Rotary Breaker Lockout Polypropylene
Catalog No. 17033786 Shop All Brady Corporation Products
Change view
Click to view available options
Product Type:
Rotary Breaker Lockout
Catalog No. Product Type
17-033-786 Rotary Breaker Lockout
1 options
Catalog No. 17-033-786 Supplier Brady Supplier No. ROT1
Add to Cart
Add to Cart
Enhance worker safety and maximize lockout efficiency with the Universal Rotary Breaker Lockout - the most adaptable rotary switch device for streamlined safety and compliance.
  • Universal fit securely accommodates most engine switches with a panel height up to 1.78 in. (45 mm).
  • Intuitive thumbwheel design ensures fast, tool-free installation and removal
  • Industrial-grade construction withstands harsh environments
Specifications
Product Type Rotary Breaker Lockout
Color Black/Red
Material Polypropylene
Dimensions (D x W x H) 1.5 x 1.5 x 3.5 in.