Learn More
Thermo Scientific™ Universal Sample Holders for Thermo Scientific Spectronic™ IR100 FT-IR Spectrometer
Catalog No. 1420020 Shop All Thermo Scientific Products
Encompass
Change view
Click to view available options
Diameter (Metric):
13 mm
Catalog No. Diameter (Metric)
14-200-20 13 mm
1 options
This item is not returnable. View return policy
Catalog No. 14-200-20 Supplier Thermo Scientific™ Supplier No. 0016213
This item is not returnable. View return policy
Universal Sample Holders. Available in two sizes.
  • Mounts for salt plates, pellets, or films
  • Fit in standard slide-card mounts
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Spectronic IR100 FT-IR spectrometer
Type Universal Sample Holder
Diameter (Metric) 13 mm
For Use With (Application) For positioning of KBr pellets, thin films, or IR windows for transmission analyses