Learn More
Kuhner Shaker Inc UNIVERSAL SANDWICH COVER FOR 2
Catalog No. 502570152
Encompass
Universal sandwich cover for 24-well MTPs (with solid silicone, 18 mm hole distance, 2.5 mm holes)
Catalog No. 50-257-0152 Supplier Kuhner Shaker Inc Supplier No. SMCR1324B
Universal sandwich cover for 24-well MTPs (with solid silicone, 18 mm hole distance, 2.5 mm holes)