Learn More
Desco™ Universal Snap Kit
SDP
Catalog No. 18688042
Click to view available options
Color:
Yellow/Silver
Catalog No. Color
18-688-042 Yellow/Silver
1 options
Catalog No. 18-688-042 Supplier Desco™ Supplier No. 09864
Universal Snap Kit includes a 10 mm stud, 10 mm socket, long screw, and short screw.
  • Easy to instal
  • Can be used for a range of mat thickness
  • ESD common point ground symbol label
  • Used to indicate the location of an acceptable common point ground as defined by ANSI/EOS-ESD-S6.1
  • Made in The United States of America
Specifications
Color Yellow/Silver
Material Stainless Steel, Brass with nickel plating, Polycarbonate
Width (Metric) 10 mm