Learn More
AVANTOR MASTERFLEX (VWR LLC) Rubberfab WN/TR SOCKET Torque Tee Sanitary Clamp Securing Device Universal Socket
Catalog No. NC1276756
Encompass
Use the Torque Tee to install and remove sanitary clamps safely and securely, even in hard-to-reach spaces. The shape of the handle provides plenty of leverage to reach the factory preset torque. Just listen for the audible click to know the clamp wing nut is properly tightened. If you need to loosen or remove the clamp, simply disengage the ratcheting mechanism to lock the handle.
Catalog No. NC1276756 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX2350808
Use the Torque Tee to install and remove sanitary clamps safely and securely, even in hard-to-reach spaces. The shape of the handle provides plenty of leverage to reach the factory preset torque. Just listen for the audible click to know the clamp wing nut is properly tightened. If you need to loosen or remove the clamp, simply disengage the ratcheting mechanism to lock the handle.