Learn More
Waters Corp Universal Sprayer Probe Assembly
Supplier:  Waters Corp 700003890
Encompass_Preferred
Replacement Waters Universal NanoFlow Sprayer, specially designed to work with fused silica emitters on Waters NanoFlow and NanoLockSpray MS sources
Catalog No. 50-792-925