Learn More
Grainger Universal Spring Platform
Catalog No. 502749815
Encompass_Preferred
The Benchmark Scientific Shaker Accessory H1010 is a universal platform designed to fit most laboratory shakers. It measures 10 x 10 inches and is constructed from stainless steel with rubber feet to prevent sliding. The platform features a grid pattern to secure flasks and other vessels during shaking. It can accommodate up to 4 liters total volume. This sturdy shaker platform allows for even agitation and is easy to clean. With its durable construction, it provides a stable base for vessels on shakers up to 3000 RPM.
Catalog No. 50-274-9815 Supplier Grainger Supplier No. 14N130
The Benchmark Scientific Shaker Accessory H1010 is a universal platform designed to fit most laboratory shakers. It measures 10 x 10 inches and is constructed from stainless steel with rubber feet to prevent sliding. The platform features a grid pattern to secure flasks and other vessels during shaking. It can accommodate up to 4 liters total volume. This sturdy shaker platform allows for even agitation and is easy to clean. With its durable construction, it provides a stable base for vessels on shakers up to 3000 RPM.