Learn More
Grainger Universal Spring Platform
Catalog No. 502751784
Encompass_Preferred
The Grainger 14N134 is a Westward brand 1/4-inch drive socket set. This 108-piece socket set includes both SAE and metric sockets ranging from 1/4 inch drive to 3/4 inch drive. The set comes organized in a sturdy plastic case for convenient storage and portability. Key features of this socket set include the chrome vanadium steel construction for durability and longevity, a knurled design on the tools for grip, and a comprehensive assortment of the most commonly used socket sizes for automotive and machinery repairs and maintenance. This professional-grade socket set provides users with the essential tools needed for a wide variety of fastening and repair jobs.
Catalog No. 50-275-1784 Supplier Grainger Supplier No. 14N134
The Grainger 14N134 is a Westward brand 1/4-inch drive socket set. This 108-piece socket set includes both SAE and metric sockets ranging from 1/4 inch drive to 3/4 inch drive. The set comes organized in a sturdy plastic case for convenient storage and portability. Key features of this socket set include the chrome vanadium steel construction for durability and longevity, a knurled design on the tools for grip, and a comprehensive assortment of the most commonly used socket sizes for automotive and machinery repairs and maintenance. This professional-grade socket set provides users with the essential tools needed for a wide variety of fastening and repair jobs.