Learn More
Bio Rad Laboratories SsoAdvanced Universal SYBR Green Supermix, 1,000 x 20 uL rxns, 10 mL
Catalog No. NC0960671
Encompass
10 ml (10 x 1 ml vials), 2x qPCR mix, contains dNTPs, Sso7d fusion polymerase, MgCl2, SYBR Green I, ROX normalization dyes, for 1,000 x 20 ul reactions.
Catalog No. NC0960671 Supplier Bio Rad Laboratories Supplier No. 1725272
10 ml (10 x 1 ml vials), 2x qPCR mix, contains dNTPs, Sso7d fusion polymerase, MgCl2, SYBR Green I, ROX normalization dyes, for 1,000 x 20 ul reactions.