Learn More
Bio Rad Laboratories SsoAdvanced Universal SYBR Green Supermix, 2x concentrated liquid, 25 ml (5 x 5 ml vials), 2,500 x 20 μl reactions, SYBR Green I
Catalog No. NC3653580
Encompass
SsoAdvanced Universal SYBR Green Supermix, 2x concentrated liquid, 25 ml (5 x 5 ml vials), 2,500 x 20 μl reactions, SYBR Green I
Catalog No. NC3653580 Supplier Bio Rad Laboratories Supplier No. 1725274
SsoAdvanced Universal SYBR Green Supermix, 2x concentrated liquid, 25 ml (5 x 5 ml vials), 2,500 x 20 μl reactions, SYBR Green I