Learn More
Enzo Life Sciences MULTIVIEW Universal Tissue Microarray (50slides)
SDP
Catalog No. 502007113
Encompass_Preferred
MULTIVIEW Universal Tissue Microarray with 11 different human tissue specimens for a broad-spectrum of immunohistochemical staining procedures. The MULTIVIEW Universal Tissue Microarray consists of 11 human tissue specimens, including placenta, liver, tonsil, colon, skin, brain, breast (fibroadenoma), prostate, thyroid, kidney and fallopian tube. The samples are fixed in formalin and paraffin embedded, and the cores are approximately 2.0mm in diameter. The sections are cut at 3µm and placed on a SuperFrost Plus slide. Alternative name: TMA. Long Term Storage: Ambient. Handling: To preserve reactivity with some stains, slides should be stained within 12 weeks of sectioning when stored at room temperature (20-25 °C). Applications: IHC ISH (in situ hybridization).
Catalog No. 50-200-7113 Supplier Enzo Life Sciences Supplier No. ADI9500200050
MULTIVIEW Universal Tissue Microarray with 11 different human tissue specimens for a broad-spectrum of immunohistochemical staining procedures. The MULTIVIEW Universal Tissue Microarray consists of 11 human tissue specimens, including placenta, liver, tonsil, colon, skin, brain, breast (fibroadenoma), prostate, thyroid, kidney and fallopian tube. The samples are fixed in formalin and paraffin embedded, and the cores are approximately 2.0mm in diameter. The sections are cut at 3µm and placed on a SuperFrost Plus slide. Alternative name: TMA. Long Term Storage: Ambient. Handling: To preserve reactivity with some stains, slides should be stained within 12 weeks of sectioning when stored at room temperature (20-25 °C). Applications: IHC ISH (in situ hybridization).