Learn More
Perkin Elmer US LLC Universal Transmission Holder
Catalog No. 502113617
Encompass_Preferred
This transmission holder fits into the sample slide holder of a spectrometer's standard sample compartment.
Catalog No. 50-211-3617 Supplier Perkin Elmer US LLC Supplier No. L1272269
This transmission holder fits into the sample slide holder of a spectrometer's standard sample compartment.