Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copan Diagnostics Universal Transport Medium (UTM-RT) In Screw-Cap Tube
SDP

Catalog No. 21377172 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Quantity:
50 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-377-172 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-377-172 Supplier Copan Diagnostics Supplier No. 3C098N

missing translation for 'validMainframeMsg'

Ready-to-use screw-cap tube, sterile, and prelabeled for upper respiratory molecular assays. Ensures safe and optimized diagnostic accuracy

  • Copan UTM-RT™ is a ready-to-use system that requires no further preparation. It consists of a 15.5 x 95 mm screw cap tube, round bottom, containing 3 mL of light orange-red transport medium
  • UTM™ tubes are supplied individually (in bulk) or in kit format
  • UTM-RT™ is designed for direct loading on Roche cobas™ 5800/6800/8800, but also compatible and validated with all molecular platforms
  • UTM-RT™ tubes are packaged 50 tubes p/pk, 6 packs (300 tubes) p/box

Specifications

Product Type Transport Tube
Format Tube
Media Type UTM-RT
Sterility Sterile
Closure Color Light Blue
Closure Type Cap
Color Orange
Diameter (Metric) 19 to 0.20 mm
For Use With (Application) Specimen Collection and Transport
For Use With (Equipment) Roche cobas 5800/6800/8800
Includes None
Length (Metric) 15.5 x 95 mm
Packaging Bulk
pH Neutral
Quantity 50 Pk.
Storage Requirements 2 to 25°C Do Not Freeze
Transport Temperature 2 to 25°C Do Not Freeze
Tube Size 15.5 x 95 mm
Closure Material Polythelene
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.